Learn More
MSC Akro-Mils Hopper Shelf Bin, High-Density Polypropylene, 17-7/8 in. x 4-1/8 in. x 4 in., Yellow, 1 ea
Catalog No. 5000547974
Encompass
This high-density polypropylene hopper shelf bin provides a durable and efficient storage solution for industrial and commercial environments. The bin features a sturdy, one-piece construction that is resistant to water, rust, and corrosion, making it an excellent alternative to standard cardboard containers.
  • Constructed from durable industrial-grade polymers that are unaffected by grease or oil
  • Features a curved-bottom hopper front and a large front label slot for easy access and organization
  • Includes a full-width hanger lip, extra-wide stacking ledge, and reinforced side ribs for structural integrity
  • Works in temperatures ranging from -20°F to 250°F and is autoclavable
  • Fits standard wire or steel shelving
Catalog No. 50-005-47974 Supplier MSC Supplier No. 55954572
This high-density polypropylene hopper shelf bin provides a durable and efficient storage solution for industrial and commercial environments. The bin features a sturdy, one-piece construction that is resistant to water, rust, and corrosion, making it an excellent alternative to standard cardboard containers.
  • Constructed from durable industrial-grade polymers that are unaffected by grease or oil
  • Features a curved-bottom hopper front and a large front label slot for easy access and organization
  • Includes a full-width hanger lip, extra-wide stacking ledge, and reinforced side ribs for structural integrity
  • Works in temperatures ranging from -20°F to 250°F and is autoclavable
  • Fits standard wire or steel shelving