Learn More
MSC Akro-Mils Shelf Bin, Polymer, Hopper Front, 11-1/8 in. x 11-5/8 in. x 4 in., 12/Ca.
Catalog No. 5000508856
Encompass
This industrial-grade polymer shelf bin is designed for enhanced organization and storage efficiency. Featuring a durable one-piece construction, it is engineered to withstand heavy loads without bulging or cracking, providing a reliable solution for managing tools and small parts in professional environments.
  • Fits on standard shelving units
  • Built-in rear hang lock to prevent accidental spills
  • Hopper front design for easy access to contents
  • Waterproof and corrosion-proof construction
  • Resistant to grease, oil, weak acids, and alkalis
  • Nesting capability for compact storage when not in use
  • Load capacity of 20 lbs.
  • Dimensions: 11-1/8 in. W x 11-5/8 in. L x 4 in. H
Catalog No. 50-005-08856 Supplier MSC Supplier No. 43375997
This industrial-grade polymer shelf bin is designed for enhanced organization and storage efficiency. Featuring a durable one-piece construction, it is engineered to withstand heavy loads without bulging or cracking, providing a reliable solution for managing tools and small parts in professional environments.
  • Fits on standard shelving units
  • Built-in rear hang lock to prevent accidental spills
  • Hopper front design for easy access to contents
  • Waterproof and corrosion-proof construction
  • Resistant to grease, oil, weak acids, and alkalis
  • Nesting capability for compact storage when not in use
  • Load capacity of 20 lbs.
  • Dimensions: 11-1/8 in. W x 11-5/8 in. L x 4 in. H