Learn More
MSC Vikan ColorCore Nail brush, 3.7 in, Medium, Polypropylene, 1 ea
Catalog No. 502868076
Encompass
This Vikan ColorCore Nail brush is designed for effective handwashing in environments like food service and food processing. Its medium polypropylene bristles efficiently remove various soils as part of proper hand washing practices.
  • Medium bristle stiffness
  • Polypropylene bristles and plastic block material
  • Product dimensions: 3.7 in length, 1.4 in width, 2.2 in height
  • Rectangular brush head
  • Short handle
  • FDA compliant
  • Manufactured in Denmark
Catalog No. 50-286-8076 Supplier MSC Supplier No. 14396824
This Vikan ColorCore Nail brush is designed for effective handwashing in environments like food service and food processing. Its medium polypropylene bristles efficiently remove various soils as part of proper hand washing practices.
  • Medium bristle stiffness
  • Polypropylene bristles and plastic block material
  • Product dimensions: 3.7 in length, 1.4 in width, 2.2 in height
  • Rectangular brush head
  • Short handle
  • FDA compliant
  • Manufactured in Denmark