Learn More
MSC DuPont Nomex IIIA Coverall, Flame Resistant, 4.5 oz, Red, 1 ea
Catalog No. 5000508600
Encompass
This Nomex® IIIA coverall provides reliable protection against flash and pool fire hazards in industrial environments. Designed for superior thermal performance, the fabric expands when exposed to heat to form a stable, inert barrier that minimizes break-open, helping to safeguard the wearer. It is constructed from a high-strength aramid fiber blend that is permanently flame resistant and features reinforced stress points for increased durability.
  • Inherently flame resistant and anti-static properties
  • Designed to not melt, drip, or burn in air
  • Features an elastic waistband and deep-pleated action back for improved mobility
  • Includes seven double-stitched pockets for convenient storage
  • Two-way concealed brass break-away zipper closure
  • Meets NFPA 2112 and 70E standards
Catalog No. 50-005-08600 Supplier MSC Supplier No. 43384882
This Nomex® IIIA coverall provides reliable protection against flash and pool fire hazards in industrial environments. Designed for superior thermal performance, the fabric expands when exposed to heat to form a stable, inert barrier that minimizes break-open, helping to safeguard the wearer. It is constructed from a high-strength aramid fiber blend that is permanently flame resistant and features reinforced stress points for increased durability.
  • Inherently flame resistant and anti-static properties
  • Designed to not melt, drip, or burn in air
  • Features an elastic waistband and deep-pleated action back for improved mobility
  • Includes seven double-stitched pockets for convenient storage
  • Two-way concealed brass break-away zipper closure
  • Meets NFPA 2112 and 70E standards