Learn More
MSC Cortech USA Institutional Plastic Can, 2-1/2 gal, Rectangular, Gray
Catalog No. 502837660
Encompass
Cortech USA correctional facility trash can is a rectangular institutional plastic can with a 2-1/2 gallon capacity. It is designed for high-speed applications and features a durable, corrosion-resistant plastic construction. This can includes a plastic liner and is flame/fire-retardant, passing CAL-133 standards, making it suitable for secure environments.
  • Designed for institutional use
  • Features a 360 degree molded handle
  • Includes molded in floor glides
  • Comes with a plastic liner
  • Corrosion resistant
  • Flame and fire-retardant, passing CAL-133 standards
Catalog No. 50-283-7660 Supplier MSC Supplier No. 84439520
Cortech USA correctional facility trash can is a rectangular institutional plastic can with a 2-1/2 gallon capacity. It is designed for high-speed applications and features a durable, corrosion-resistant plastic construction. This can includes a plastic liner and is flame/fire-retardant, passing CAL-133 standards, making it suitable for secure environments.
  • Designed for institutional use
  • Features a 360 degree molded handle
  • Includes molded in floor glides
  • Comes with a plastic liner
  • Corrosion resistant
  • Flame and fire-retardant, passing CAL-133 standards