Learn More
MSC Vestil Polyethylene Drum Recycling Lid, Flaps, 30 gal, 1 ea
Catalog No. 502817307
Encompass
This recycling lid is constructed from low-density polyethylene, designed for collecting aluminum cans or plastic bottles. It fits securely on open-head plastic or steel drums when the original lid and ring are removed, making it suitable for outdoor environments.
  • Fits snug to resist blowing off or tampering by people or animals
  • Features a flap to discourage removal of contents
  • Strong construction durability
  • Lightweight design for simple transferring
  • Designed for use with 30 gal drums
Catalog No. 50-281-7307 Supplier MSC Supplier No. 15381379
This recycling lid is constructed from low-density polyethylene, designed for collecting aluminum cans or plastic bottles. It fits securely on open-head plastic or steel drums when the original lid and ring are removed, making it suitable for outdoor environments.
  • Fits snug to resist blowing off or tampering by people or animals
  • Features a flap to discourage removal of contents
  • Strong construction durability
  • Lightweight design for simple transferring
  • Designed for use with 30 gal drums