Learn More
Research Products International Corp Polypropylene Replacement Cap for Glass Coplin Jar
SDP
Catalog No. 50998431
Encompass
White polypropylene screw cap.  Polyethylene lid is vented for use in microwave applications. Internal grooves hold up to ten standard 1'' x 3'' slides.
Catalog No. 50-998-431 Supplier Research Products International Corp Supplier No. 144212
White polypropylene screw cap.  Polyethylene lid is vented for use in microwave applications. Internal grooves hold up to ten standard 1'' x 3'' slides.