Learn More
MSC Cortech USA Trash Can, 30 gal, Polyethylene, Round
Catalog No. 502837664
Encompass
This round polyethylene trash can has a 30-gallon capacity, designed for durability and ease of use in various environments. It features 360-degree molded-in handles and molded-in floor glides to reduce scratching. The can is virtually unbreakable and an optional food-grade version is available.
  • Option for flame retardant or food grade material.
  • 360-degree molded-in continuous handles surround the entire can perimeter and underside for easier lifting.
  • Molded-in floor glides reduce floor scratching when dragged across the floor.
  • Virtually unbreakable.
  • Manufactured to ISO quality standards in the USA.
Catalog No. 50-283-7664 Supplier MSC Supplier No. 84440973
This round polyethylene trash can has a 30-gallon capacity, designed for durability and ease of use in various environments. It features 360-degree molded-in handles and molded-in floor glides to reduce scratching. The can is virtually unbreakable and an optional food-grade version is available.
  • Option for flame retardant or food grade material.
  • 360-degree molded-in continuous handles surround the entire can perimeter and underside for easier lifting.
  • Molded-in floor glides reduce floor scratching when dragged across the floor.
  • Virtually unbreakable.
  • Manufactured to ISO quality standards in the USA.