Learn More
MSC Cortech USA Cobra Can, 22 Gallon, Food Grade/General Use, Structural Polyethylene
Catalog No. 502839066
Encompass
The Cobra Can waste container is constructed from virtually unbreakable, rotationally molded structural polyethylene, designed for continuous and intensive use in demanding environments. This durable container is suitable for both food grade and general use applications.
  • Virtually unbreakable
  • 360° molded-in continuous handles for easier lifting
  • Molded-in floor glides reduce floor scratching
  • .22" Nominal wall thickness
  • Easy to clean, sanitize, and maintain
  • Manufactured to ISO quality standards in the USA
  • 10 year factory warranty on manufacturing defects
Catalog No. 50-283-9066 Supplier MSC Supplier No. 84439850
The Cobra Can waste container is constructed from virtually unbreakable, rotationally molded structural polyethylene, designed for continuous and intensive use in demanding environments. This durable container is suitable for both food grade and general use applications.
  • Virtually unbreakable
  • 360° molded-in continuous handles for easier lifting
  • Molded-in floor glides reduce floor scratching
  • .22" Nominal wall thickness
  • Easy to clean, sanitize, and maintain
  • Manufactured to ISO quality standards in the USA
  • 10 year factory warranty on manufacturing defects