Learn More
MSC Cortech USA Trash Can, 45 Gallon, Round, Polyethylene, Blue
Catalog No. 502839540
Encompass
The Cobra Can is a heavy-duty, virtually indestructible trash receptacle. It features 360° molded-in continuous handles and molded-in floor glides, designed to withstand extreme conditions and suitable for various facility maintenance needs.
  • Option for flame retardant or food grade material
  • 360° molded-in continuous handles for easier lifting
  • Molded-in floor glides reduce floor scratching
  • Virtually unbreakable design
  • Flame retardant version passes Cal 133
  • 22-inch nominal wall thickness
  • Can dolly sold separately
  • Manufactured to ISO quality standards in the USA
Catalog No. 50-283-9540 Supplier MSC Supplier No. 84442482
The Cobra Can is a heavy-duty, virtually indestructible trash receptacle. It features 360° molded-in continuous handles and molded-in floor glides, designed to withstand extreme conditions and suitable for various facility maintenance needs.
  • Option for flame retardant or food grade material
  • 360° molded-in continuous handles for easier lifting
  • Molded-in floor glides reduce floor scratching
  • Virtually unbreakable design
  • Flame retardant version passes Cal 133
  • 22-inch nominal wall thickness
  • Can dolly sold separately
  • Manufactured to ISO quality standards in the USA