Learn More
StatLab Poly Scientific Ammoniacal Silver Nitrate Solution - Gomoris

Description
- Used to demonstrate reticulum fibers in tissue, it is part of Gomori's method for reticulum fibers, which involves a series of steps to prepare the tissue for staining
- The solution is used to create submicroscopic sensitized sites of silver that are then deposited as dark brown or black deposits on the reticular fibers.
- A StatLab - Poly Scientific Special Stain Kit Component
Specifications
Specifications
| Physical Form | Solution |
| CAS | 1310-58-3, 1336-21-6, 7761-88-8 |
| Color | Colorless |
| Quantity | 1 gal. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.