Learn More
StatLab Poly Scientific Azure B Counterstain For Melanin Pigment
Description
- Allows pathologists to interpret immunohistologic preparations of melanocytic lesions effectively
- The stain is particularly useful in heavily pigmented lesions, such as melanomas, blue nevi, and pigmented spindle-cell nevi
- By metachromatically staining melanin granules, Azure B helps in distinguishing these lesions from other types of tissue that may appear similar
- This distinction is crucial for accurate diagnosis and treatment planning in dermatology and other fields of pathology
Specifications
Specifications
| Product Type | Azure B Counterstain |