Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Basic Fuchsin 0.5% Aqueous
SDP

Catalog No. 21010349
Change view
Click to view available options
Product Type:
Basic Fuchsin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
21-010-349 Basic Fuchsin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-349 Supplier StatLab Supplier No. S12216OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Used in the Gram stain to identify gram negative bacteria.

  • A StatLab - Poly Scientific Special Stain Kit Component
  • Ideal for staining Gram-negative bacteria

Specifications

Quantity 16 oz.
Volume (Metric) 473 mL
Product Type Basic Fuchsin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.