Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Buffered Formic Acid Trisodium Citrate Formic Acid
SDP

Catalog No. 23640207
Change view
Click to view available options
Color:
Colorless
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-207 Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-640-207 Supplier StatLab Supplier No. S169K4X1GL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to help in the decalcification process, which is crucial for preparing high-quality paraffin sections from bone or other calcified tissues

  • Solution is effective for routine histological specimens and is available in quantities of four 1-gallon rectangular F-style containers
  • This product is specifically designed to help in the decalcification process, which is crucial for preparing high-quality paraffin sections from bone or other calcified tissues
  • The decalcification process is carried out after the specimen has been thoroughly fixed and prior to routine processing to paraffin
  • A StatLab - Poly Scientific Special Stain Kit Component
  • 4 Gallons per Case

Specifications

Physical Form Solution
CAS 64-18-6, 6132-04-3
Color Colorless
Quantity 1 gal.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.