Learn More
StatLab Poly Scientific Carbol Fuchsin Kinyoun's Stain
SDP
Catalog No. 22023775
Change view
Click to view available options
Quantity:
16 oz.
Catalog No. Quantity
22-023-775 16 oz.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-775 Supplier StatLab Supplier No. S16116OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab Poly Scientific Carbol Fuchsin Kinyoun's Stain - used to identify acid-fast bacteria, particularly Mycobacterium tuberculosis
  • This stain works by penetrating the waxy cell walls of acid-fast bacteria, which contain mycolic acids, making them resistant to decolorization
  • The Kinyoun stain is particularly useful because it does not require heating, unlike the Ziehl-Neelsen method, and allows for the differentiation between acid-fast and non-acid-fast organisms.
  • A StatLab - Poly Scientific Special Stain Kit Component
Specifications
Quantity 16 oz.
Product Type Carbol Fuchsin