Learn More
StatLab Poly Scientific Carbol Fuchsin Ziehl Neelsen Stain
SDP
Catalog No. 22023782
Change view
Click to view available options
Quantity:
8 oz.
16 oz.
32 oz.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-782 16 oz.
22-023-774 32 oz.
22-023-777 8 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-782 Supplier StatLab Supplier No. S16216OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab Poly Scientific Carbol Fuchsin Ziehl Neelsen Stain - used for the qualitative demonstration of acid fast organisms
  • Used to identify acid-fast bacteria, particularly those belonging to the Mycobacterium genus, which includes the causative agent of tuberculosis (Mycobacterium tuberculosis) and leprosy (Mycobacterium leprae)
  • A StatLab - Poly Scientific Special Stain Kit Component
Specifications
Quantity 16 oz.
Product Type Carbol Fuchsin