Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Cellosolve
SDP

Catalog No. 22023761
Change view
Click to view available options
Quantity:
32 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-761 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-761 Supplier StatLab Supplier No. S163D32OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

StatLab Poly Scientific Cellosolve - used for dehydration and preservation of tissue samples

  • Allows for rapid dehydration of tissues by transferring them from water or other solutions directly to a 2% solution of celloidin in Cellosolve, which helps maintain tissue features without hardening
  • Cellosolve can enhance the visibility of Gram-negative bacteria by removing excess fuchsine from cytoplasmic elements in the background
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Product Type Dehydrant 100%
Quantity 32 oz.
Solution Type Stain Kit
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.