Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Chromic Acid 4% Aqueous Fixative
SDP

Catalog No. 22023778
Change view
Click to view available options
Quantity:
8 oz.
16 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-778 8 oz.
22-023-772 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-778 Supplier StatLab Supplier No. S1648OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Poly Scientific Chromic Acid 4% Aqueous Fixative

  • It acts by oxidizing and cross-linking proteins, stabilizing cell structures, and preserving them for later examination under a microscope
  • This preservation of morphology allows for accurate assessment of cell structures
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Physical Form Solution
Color Red
Quantity 8 oz.
Concentration 4%
Grade Histological
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.