Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Chromic Acid 5% Aqueous
SDP

Catalog No. 23640011
Change view
Click to view available options
Quantity:
8 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-011 8 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-011 Supplier StatLab Supplier No. S1658OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

Used in the Gomori’s methenamine staining procedure for effective detection of fungi in tissue sections.

  • Used as part of the Grocott-Gomori methenamine stain (GMS) technique, a widely established method in microbiology and histopathology for identifying fungal infections in tissue samples.
  • The staining procedure utilises a 5% w/v chromic acid solution to enhance the visibility and contrast of fungal structures, supporting accurate microscopic identification, diagnosis, and appropriate clinical treatment.
  • Supplied as a StatLab Poly Scientific Special Stain Kit component, ensuring consistent formulation, compatibility, and reliable performance in professional laboratory staining workflows.

Specifications

Quantity 8 oz.
Product Type Stain Kit Component
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.