Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Crystal Violet 1% Aqueous
SDP

Catalog No. 21010350
Change view
Click to view available options
Product Type:
Crystal Violet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
21-010-350 Crystal Violet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-350 Supplier StatLab Supplier No. S167D8OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Used in gram stains for the staining of bacteria.

  • The primary stain in the Gram staining method, which differentiates Gram-positive and Gram-Negative bacteria
  • The dye infiltrates the peptidoglycan-rich cell wall of Gram-positive bacteria and forms entrapped complexes with Gram's iodine, retaining the purple color post-decolorization
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Quantity 8 oz.
Volume (Metric) 237 mL
Product Type Crystal Violet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.