Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Davidson's Fixative Blue
SDP

Catalog No. 23800517
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-800-517 5 gal.
23-800-516 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-800-517 Supplier StatLab Supplier No. S22505GL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rapid fixative with good nuclear detail, useful for preparing tumors, bone marrow specimens, gynecologic material, fatty breast, and medical biopsies

  • It is particularly useful for preparing tumors, bone marrow specimens, gynecologic material, fatty breast, and medical biopsies
  • Solution is particularly beneficial for testicular biopsies and for overnight fixation of fat-containing lymph nodes
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Quantity 5 gal.
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.