Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Davidson's Fixative Modified
SDP

Catalog No. 22021457
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
22-021-457 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-457 Supplier StatLab Supplier No. S28031GL328250
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Used for the preservation of various tissues for examination.

  • It is particularly useful for the following:
  • Tumors: Provides good nuclear detail with minimal formalin pigment
  • Bone Marrow: Ideal for preparing specimens for histopathological examination
  • Gynecologic Material: Useful for examining gynecologic specimens
  • Fatty Breast: Helps in the examination of fatty breast tissues
  • Medical Biopsies: Assists in the examination of medical biopsies
  • Testicular Biopsies: Used for the fixation of testicular specimens
  • Lymph Nodes: Effective for the visualization of lymph nodes in radical dissection specimens
  • Eye Fixation: Recommended for eye fixation to support excellent staining
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Quantity 1 gal.
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.