Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Davidson's Fixative Pink
SDP

Catalog No. 23640014
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-014 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-014 Supplier StatLab Supplier No. S22501GL514300
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Rapid fixative used to preserve tissue samples for histological examination.

  • Rapid-acting Davidson’s Fixative solution suitable for preserving a wide range of tissue samples, including tumors, bone marrow specimens, gynecologic material, fatty breast tissue, and medical biopsy samples.
  • Provides excellent nuclear detail with minimal formalin pigment formation, making it ideal for tissues that are easily distorted during processing.
  • Distinctive pink-colored solution allows for easy visual identification and handling in the laboratory environment.
  • Supplied as a StatLab Poly Scientific Special Stain Kit component, ensuring consistent quality, compatibility, and reliable performance in histology workflows.

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Quantity 1 gal.
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.