Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Davidsons Fixative Alternate with Blue Tint
SDP

Catalog No. 21010356
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-010-356 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-356 Supplier StatLab Supplier No. S26564X1GL514300
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Used for its ability to preserve tissue while minimizing shrinkage and maintaining good nuclear detail.

  • It is particularly useful for preparing tumors, bone marrow specimens, gynecologic material, fatty breast, and medical biopsies
  • Solution is also known for its effectiveness in preparing whole eyes, which helps avoid retinal detachment during fixation
  • The fixative is non-toxic and does not contain mercury or other metals, making it a safer alternative for histological procedures
  • 4/Cs.
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Quantity 1 gal.
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.