Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Davidsons Fixative Modified
SDP

Catalog No. 22023757
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-757 5 gal.
22-023-756 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-757 Supplier StatLab Supplier No. S28035GL328250
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Poly Scientific Davidsons Fixative Modified - a rapid fixative with good nuclear detail

  • A histological fixative used for the preservation of various tissues for examination
  • It is particularly useful for tumors, bone marrow, gynelogical specimens, fatty breast biopsies, testicular biopsies and lymph node specimens.
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Fixative
Quantity 5 gal.
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.