Learn More
StatLab Poly Scientific EA-36
SDP
Catalog No. 23640024
Change view
Click to view available options
Quantity:
4 x 1 gal.
Catalog No. Quantity
23-640-024 4 x 1 gal.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-024 Supplier StatLab Supplier No. S2504X1GL
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Cytoplasmic counterstain used in histology to provide brilliant cytoplasmic staining for gynecological and non-gynecological specimens.
  • EA-36 cytoplasmic counterstain formulated for use in histology and cytology, delivering brilliant and consistent cytoplasmic staining in both gynecological and non-gynecological specimens.
  • Commonly used in Papanicolaou staining methods, where it assists in differentiating cells in smear preparations from a variety of bodily secretions.
  • Known for staining the cytoplasm of all other cells to produce a characteristic multicolored effect, which is essential for accurate interpretation of cytological specimens.
  • Supplied as a StatLab Poly Scientific Special Stain Kit component, ensuring reliable performance, consistency, and compatibility in laboratory staining workflows.
Specifications
Quantity 4 x 1 gal.
Product Type Counterstain