Learn More
StatLab Poly Scientific Fontana Masson Silver Method For Argentaffin Granules Stain Kit

Description
- The Fontana Masson stain is used in histology lab applications to demonstrate argentaffinic substances, including melanin and argentaffin granules
- It is useful in cases of carcinoid tumors and melanoma
- The stain identifies silver-reducing argentaffin molecules in tissue sections
Specifications
Specifications
| Physical Form | Aqueous Solution |
| Color | Clear Colorless |
| Quantity | 4 oz. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.