Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Fontana Masson Silver Method For Argentaffin Granules Stain Kit
SDP

Catalog No. 23640181
Change view
Click to view available options
Quantity:
4 oz.
8 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-181 8 oz.
23-640-180 4 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-181 Supplier StatLab Supplier No. K0128OZ
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Fontana Masson Silver Method For Argentaffin Granules

  • The Fontana Masson stain is used in histology lab applications to demonstrate argentaffinic substances, including melanin and argentaffin granules
  • It is useful in cases of carcinoid tumors and melanoma
  • The stain identifies silver-reducing argentaffin molecules in tissue sections

Specifications

Physical Form Aqueous Solution
Color Clear Colorless
Quantity 8 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.