Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Formalin 20% Aqueous
SDP

Catalog No. 21010351
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-010-351 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-351 Supplier StatLab Supplier No. S18078OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

A general purpose fixative for all tissue to be examined by light microscopy.

  • Fixative is particularly effective for surgical and necropsy specimens and is an excellent fixative for brain tissue
  • This fixative is essential for preserving tissue samples for further analysis and research purposes
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Quantity 8 oz.
Solution Type Formalin
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.