Learn More
StatLab Poly Scientific Giemsa Stain Stock Solution
SDP
Catalog No. 22021461
Change view
Click to view available options
Quantity:
32 oz.
Catalog No. Quantity
22-021-461 32 oz.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-461 Supplier StatLab Supplier No. S19532OZ
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Use with Giemsa Stain Kits for the demonstration of bone marrow, mast cells, H. pylori, and other hematopoietic tissues.
  • Giemsa stain stock solution is primarily used for the visualization of cellular components, especially in blood smears and bone marrow samples
  • It is a classic histological stain used for the visualization of cellular components, especially in blood smears and bone marrow samples
  • It is widely used in the diagnosis of various blood disorders and in the detection of parasites such as Plasmodium - the causative agent of malaria
  • A StatLab - Poly Scientific Special Stain Kit Component
Specifications
Quantity 32 oz.
Product Type Giemsa Stain