Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Giemsa Stain Stock Solution
SDP

Catalog No. 22023776
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-776 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-776 Supplier StatLab Supplier No. S19516OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Poly Scientific Giemsa Stain Stock Solution - for the demonstration of bone marrow, mast cells, H. pylori, and other hematopoietic tissues

  • Giemsa stock solution is primarily used for the differential staining of blood cells
  • It helps in differentiating between human and bacterial cells, with human cells appearing purple and bacterial cells appearing pink
  • This staining is crucial for the identification of various blood parasites and for the morphological examination of blood cells such as erythrocytes, leukocytes, and platelets
  • The Giemsa stain is also used in cytogenetics to stain chromosomes and identify chromosomal aberrations
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Physical Form Solution
Color Pink
Quantity 16 oz.
Grade Histological
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.