Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Glutaraldehyde 0.625% In Phosphate Buffer pH 7.4 0
SDP

Catalog No. 23640023
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-023 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-023 Supplier StatLab Supplier No. S223616OZ
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Used as a fixative for electron microscopy sample preparation.

  • Ready-to-use glutaraldehyde 0.625% solution in phosphate buffer pH 7.4, formulated to maintain stable pH during fixation and preserve the structural integrity of biological samples.
  • Pre-mixed buffered formulation offers convenience for routine quality control processes and may be further diluted with distilled water when required.
  • Essential fixative for electron microscopy, supporting optimal preservation of cellular ultrastructure for detailed microscopic examination and analysis.
  • Supplied as a StatLab Poly Scientific Special Stain Kit component, ensuring consistency, compatibility, and reliable laboratory performance.

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixation Buffer
Quantity 16 oz.
Solution Type Buffer
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.