Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Glutaraldehyde 3% In Sorensen's Phosphate Buffer pH 7.2
SDP

Catalog No. 21010353
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-010-353 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-353 Supplier StatLab Supplier No. S1978OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Used as a fixative to preserve tissue samples for electron microscopy.

  • Solution is effective in fixing tissues by penetrating the specimen and allowing the fixative to react with the tissue elements, stabilizing them in place
  • This fixative is particularly useful in electron microscopy studies, where the preservation of tissue samples is crucial for detailed analysis
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Solution Type Fixation Buffer
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.