Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Gold Chloride 0.2% Aqueous
SDP

Catalog No. 22021452
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-452 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-452 Supplier StatLab Supplier No. S20216OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

Used as a toner by replacing some of the bound silver with gold.

  • Used as a reducing agent for staining and visualizing neurons and nerve fibers
  • It is utilized in various silver plating methods to visualize neuronal structures such as dendrites and axons
  • The solution binds to and reduces silver compounds applied to tissue specimens, resulting in visible silver grains that adhere to neuronal structures
  • Can be used with Grocott's Method for Fungi (GMS)
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Quantity 16 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.