Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Gomori's Methenamine Silver Nitrate
SDP

Catalog No. 22033325
Change view
Click to view available options
Color:
Colorless
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-033-325 Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-033-325 Supplier StatLab Supplier No. K0168OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fungi and pneumocystis carinii special stain kit, used for the identification carinii in tissue

  • Intended for the qualitative demonstration of fungi and Pneumocysistis carinii in tissue
  • Each kit consists of Chromic Acid 5%, Borax 5% Aqueous, Methenamine Silver Nitrate Stock Solution, Sodium Metabisulfite 1% Aqueous, 0.2% Gold Chloride Aqueous, Sodium Thiosulfate 2% Aqueous, and Fast Green Substitute
  • All kit components are available for purchase individually
  • Please review all component labels to ensure correct storage temperature
  • A StatLab - Poly Scientific Special Stain Kit

Specifications

Physical Form Solution
CAS 1303-96-4
Color Colorless
Quantity 4 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.