Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Gomori's Method
SDP

Catalog No. 22033326
Change view
Click to view available options
Quantity:
8 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-033-326 8 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-033-326 Supplier StatLab Supplier No. K0188OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to visualize reticular fibers, which are a type of connective tissue composed of type III collagen

  • These fibers are particularly useful in the study of lymphatic tissues and bone marrow
  • The method involves a series of steps to impregnate silver into the reticular fibers, creating a dark brown or black deposit that can be observed under a microscope
  • This technique is essential for researchers and pathologists who need to analyze the structure and function of reticular fibers in various tissues
  • A StatLab - Poly Scientific Special Stain Kit

Specifications

Physical Form Solution
CAS 7761-88-8, 1310-58-3, 1336-21-6
Color Colorless
Quantity 8 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.