Learn More
StatLab Poly Scientific Grocott's Method for Fungi, Microwave Stain Kit
SDP
Catalog No. 22023767
Change view
Click to view available options
Quantity:
8 oz.
Catalog No. Quantity
22-023-767 8 oz.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-767 Supplier StatLab Supplier No. MW0048OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab Poly Scientific Grocott's Method For Fungi (GMS), Microwave Stain Kit - intended for the qualitative demonstration of fungi and Pneumocysistis carinii in tissue
  • Each kit consists of Chromic Acid 4% Aqueous, Methenamine Silver Nitrate Stock Solution Set, Sodium Metabisulfite 1% Aqueous, 0.1% Gold Chloride Aqueous, Sodium Thiosulfate 2% Aqueous, and Fast Green Substitute
  • Available for purchase in 8 oz. or 16 oz. bottles and all kit components are available for purchase individually
  • Please review all component labels to ensure correct storage temperature
  • A StatLab - Poly Scientific Special Stain Kit
Specifications
Quantity 8 oz.