Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Grocott's Method for Fungi Special Stain Kit
SDP

Catalog No. 22023786
Change view
Click to view available options
Quantity:
8 oz.
16 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-786 8 oz.
22-023-758 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-786 Supplier StatLab Supplier No. K0228OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Poly Scientific Grocott's Method for Fungi (GMS) Special Stain Kit - Intended for the qualitative demonstration of fungi and Pneumocysistis carinii in tissue

  • Each kit consists of Chromic Acid 4% Aqueous, Methenamine Silver Nitrate Stock Solution Set, Sodium Metabisulfite 1% Aqueous, 0.1% Gold Chloride Aqueous, Sodium Thiosulfate 2% Aqueous, and Fast Green Substitute
  • Available for purchase in 8 oz. or 16 oz. bottles
  • All kit components are available for purchase individually
  • Please review all component labels to ensure correct storage temperature
  • A StatLab Poly Scientific R and D Special Stain Kit

Specifications

Color Colorless
Form Solution
Quantity 8 oz.
Product Type Grocott's Method for Fungi Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.