Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Halt
SDP

Catalog No. 21010354
Change view
Click to view available options
Physical Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Physical Form
21-010-354 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-354 Supplier StatLab Supplier No. S243032OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Used to acclimatize tissue to the first stage of the dehydrating process.

  • This step is crucial as it prevents the drying effect on small specimens and prepares the tissue for further processing
  • The halt solution helps to acclimatize the tissue to the dehydrating process and is essential for the proper preservation and processing of histological specimens
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Physical Form Solution
Color Blue
Quantity 32 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.