Learn More
StatLab Poly Scientific HCS Calcium Control Slides
SDP
Catalog No. 21010268
Change view
Click to view available options
Quantity:
100 Slides/Box
Catalog No. Quantity
21-010-268 100 Slides/Box
1 options
Catalog No. 21-010-268 Supplier StatLab Supplier No. CS010PS100
Add to Cart
Add to Cart
It consists of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • StatLab HCS control slides are created using non-adhesion or adhesion white glass slides with 90° corners
  • Each package includes a specific staining protocol
Specifications
Quantity 100 Slides/Box