Learn More
StatLab Poly Scientific HCS PAS Control Slides
Description
- Use these slides with confidence to evaluate stain quality and performance in histology laboratories
- StatLab HCS control slides are created using non-adhesion or adhesion white glass slides with 90° corners
- Each package includes a specific staining protocol
Specifications
Specifications
| Product Type | Control Slide |
| Quantity | 25 Slides/Box |