Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Hematoxylin 5% Alcoholic
SDP

Catalog No. 22023783
Change view
Click to view available options
Quantity:
8 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-783 8 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-783 Supplier StatLab Supplier No. S212B8OZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab Poly Scientific Hematoxylin 5% Alcoholic - a key stain in the diagnosis of malignancies and other pathalogical conditions, as it provides a clear view of the cellular and tissue details

  • Used to stain the nuclei of cells, providing a clear view of the nuclear detail and chromatin structure
  • Also used for the staining of Cytoplasm: It helps in the staining of the cytoplasmic components, which are often of interest in the context of cell function and morphology
  • It is used to stain various tissues, including skin, colon, and other organs, to reveal their cellular and tissue structure
  • A StatLab - Poly Scientific Special Stain Kit Component

Specifications

Physical Form Aqueous Solution
Color Colorless
Quantity 8 oz.
Concentration 5%
Grade Histological
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.