Learn More
StatLab Poly Scientific Infiltrating/Embedding Paraffin Prills
SDP
Catalog No. 23640158
Change view
Click to view available options
Form:
Liquid
Catalog No. Form
23-640-158 Liquid
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-158 Supplier StatLab Supplier No. C8276X3LB
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Poly Scientific Infiltrating/embedding paraffin
  • Paraffin embedding involves infiltrating tissue samples with melted paraffin wax to provide a stable medium for sectioning
  • The process preserves tissue morphology and cellular structure, making it easier to cut thin sections for microscopic examination
Specifications
Product Type Paraffin
Quantity 3 lb.
Form Liquid