Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Mayer's Modified Hematoxylin
SDP

Catalog No. 23640068
Change view
Click to view available options
Color:
Purple/Violet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-068 Purple/Violet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-640-068 Supplier StatLab Supplier No. S2161GL
Add to Cart
Add to Cart

Can be used in H and E staining procedures and in many Special Stains procedures as a counterstain

  • Used for its ability to stain all cellular material, including nuclei (blue) and cytoplasmic elements (pink-red)
  • It is particularly useful in special staining techniques where a more intense nuclear staining is required
  • A StatLab - Poly Scientific H and E Stain

Specifications

Physical Form Solution
CAS 517-28-2
Color Purple/Violet
Quantity 1 gal.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.