Learn More
StatLab Poly Scientific Mayer's Mucicarmine Method

Description
- Mayers Mucicarmine Method is a histological staining technique used to identify epithelial mucins, which are secretions produced by various epithelial and connective tissue cells
- This method is particularly useful in distinguishing mucin-negative undifferentiated squamous cell lesions from mucin-positive adenocarcinomas
- It also stains the mucopolysaccharide capsule of Cryptococcus neoformans, a pathogenic fungus
- A StatLab - Poly Scientific Special Stain Kit
Specifications
Specifications
| Physical Form | Solution |
| CAS | 64-17-5, 517-28-2, 67-56-1, 67-63-0 |
| Color | Light Brown |
| Quantity | 16 oz. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.