Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Poly Scientific Mayer's Mucicarmine Method
SDP

Catalog No. 22033328
Change view
Click to view available options
Quantity:
4 oz.
16 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-033-328 4 oz.
22-033-327 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-033-328 Supplier StatLab Supplier No. K0398OZ
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Intended for the qualitative demonstration of mucin or cryptococcus

  • Mayers Mucicarmine Method is a histological staining technique used to identify epithelial mucins, which are secretions produced by various epithelial and connective tissue cells
  • This method is particularly useful in distinguishing mucin-negative undifferentiated squamous cell lesions from mucin-positive adenocarcinomas
  • It also stains the mucopolysaccharide capsule of Cryptococcus neoformans, a pathogenic fungus
  • A StatLab - Poly Scientific Special Stain Kit

Specifications

Physical Form Solution
CAS 64-17-5, 517-28-2, 67-56-1, 67-63-0
Color Light Brown
Quantity 4 oz.
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.