Learn More
StatLab Poly Scientific Mayer's Mucicarmine Solution
SDP
Catalog No. 22023785
Change view
Click to view available options
Quantity:
8 oz.
16 oz.
32 oz.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-023-785 16 oz.
22-023-766 32 oz.
22-023-773 8 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-785 Supplier StatLab Supplier No. S24616OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab Poly Scientific Mayer's Mucicarmine Solution - Effective with the staining and identifying mucins
  • Mayer's Mucicarmine Solution is a crucial stain in histology for detecting mucins, which are high molecular weight glycoproteins found in various epithelial and connective tissue cells
  • These mucins are secreted by epithelial cells in certain inflammations and in certain intestinal carcinomas
  • The solution is particularly useful in identifying mucin-positive tumor cells in areas that do not contain mucin-producing cells, indicating the tumor's origin
  • It is also effective in staining encapsulated fungi, such as cryptococcus, and is a key component in the Mayer Mucicarmine Staining Procedure
  • A StatLab - Poly Scientific Special Stain Kit Component
Specifications
Color Deep Maroon Red
Form Aqueous Solution
Quantity 16 oz.
Product Type Mucicarmine Stock Solution