Learn More
StatLab Poly Scientific Modified Gram Method For Bacteria Stain Kit
SDP
Catalog No. 21010302
Change view
Click to view available options
Volume (Metric):
237 mL
Catalog No. Volume (Metric)
21-010-302 237 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-302 Supplier StatLab Supplier No. K1068OZ
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Used to differentiate between Gram-positive and Gram-negative bacteria.
  • The kit allows for the differentiation of bacteria by their cell wall composition, which is the basis of the color affinity used in the staining process
  • Gram-positive bacteria appear blue-violet due to the murein structure of their cell wall, while Gram-negative bacteria appear red due to the re-release of the staining dye with the decolorizing solution
  • A StatLab - Poly Scientific Special Stain Kit
Specifications
Volume (Metric) 237 mL
Product Type Gram's Stain Kit