Learn More
StatLab Poly Scientific Nuclear Fast Red Kernechtrot 0.1%

Description
- It selectively stains nuclear chromatin red and provides non-specific background tissue staining in shades of pink
- This dye is particularly useful for analyzing various sample types, including hematologic elements, bone and calcium, carbohydrates and mucosubstances, connective tissue, and fats and lipids
- It is often used in conjunction with silver oxidizing solutions to enhance the differentiation of specific cellular components
- The solution is stable and can be used for a long time without significant loss of quality
- A StatLab - Poly Scientific Special Stain Kit Component
Specifications
Specifications
| Quantity | 8 oz. |
| Product Type | Nuclear Fast Red |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.