Learn More
StatLab Poly Scientific Paraformaldehyde 4% In 0.1M Phosphate Buffer pH 7.4

Description
- Solution is particularly effective in preserving the structural integrity of tissues during the fixation process
- Ideal for use in immunohistochemistry (IHC) and immunofluorescence (IF) techniques, where the fixation step is crucial for the detection of specific proteins and markers
- A StatLab - Poly Scientific Special Stain Kit Component
Specifications
Specifications
| For Use With (Equipment) | All Tissue Processors |
| Grade | Chemical Solution |
| Product Type | Paraformaldehyde |
| Solution Type | Buffer |
| Form | Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.